Merck
  • Design and synthesis of chiral N-chloroimidodicarbonates: application to asymmetric chlorination of silyl enol ethers.
Design and synthesis of chiral N-chloroimidodicarbonates: application to asymmetric chlorination of silyl enol ethers.
The Journal of organic chemistry (2007-06-05)
Saumen Hajra, Manishabrata Bhowmick, Biswajit Maji, Debarshi Sinha
ABSTRACT
New chiral N-chloroimidodicarbonates, which function as efficient chiral chlorinating agents, were designed and synthesized. Among these, C(2)-symmetric (1R,2S,5R)-(-)-menthyl-N-chloroimidodicarbonate 2a provided moderate to good enantioselectivity (up to 40%) for the chlorination of silyl enol ethers to afford alpha-chloroketones only in the presence of a suitable Lewis acid such as Sm(OTf)(3).
Sigma-Aldrich
(1R)-(−)-Menthyl chloroformate, optical purity ee: 99% (GLC)