Merck
  • Fluoride relay: a new concept for the rapid preparation of anhydrous nucleophilic fluoride salts from KF.
Fluoride relay: a new concept for the rapid preparation of anhydrous nucleophilic fluoride salts from KF.
Chemical communications (Cambridge, England) (2007-01-26)
Haoran Sun, Stephen G DiMagno
ABSTRACT
Fluoride relay is used to generate exceptionally nucleophilic fluoride reagents from KF on a time scale commensurate with radiotracer synthesis.
Sigma-Aldrich
Methyl 2,6-dichloropyridine-3-carboxylate, 97%