Merck
  • ML264, A Novel Small-Molecule Compound That Potently Inhibits Growth of Colorectal Cancer.
ML264, A Novel Small-Molecule Compound That Potently Inhibits Growth of Colorectal Cancer.
Molecular cancer therapeutics (2015-12-02)
Ainara Ruiz de Sabando, Chao Wang, Yuanjun He, Mónica García-Barros, Julie Kim, Kenneth R Shroyer, Thomas D Bannister, Vincent W Yang, Agnieszka B Bialkowska
ABSTRACT
Colorectal cancer is one of the leading causes of cancer mortality in Western civilization. Studies have shown that colorectal cancer arises as a consequence of the modification of genes that regulate important cellular functions. Deregulation of the WNT and RAS/MAPK/PI3K signaling pathways has been shown to be important in the early stages of colorectal cancer development and progression. Krüppel-like factor 5 (KLF5) is a transcription factor that is highly expressed in the proliferating intestinal crypt epithelial cells. Previously, we showed that KLF5 is a mediator of RAS/MAPK and WNT signaling pathways under homeostatic conditions and that it promotes their tumorigenic functions during the development and progression of intestinal adenomas. Recently, using an ultrahigh-throughput screening approach we identified a number of novel small molecules that have the potential to provide therapeutic benefits for colorectal cancer by targeting KLF5 expression. In the current study, we show that an improved analogue of one of these screening hits, ML264, potently inhibits proliferation of colorectal cancer cells in vitro through modifications of the cell-cycle profile. Moreover, in an established xenograft mouse model of colon cancer, we demonstrate that ML264 efficiently inhibits growth of the tumor within 5 days of treatment. We show that this effect is caused by a significant reduction in proliferation and that ML264 potently inhibits the expression of KLF5 and EGR1, a transcriptional activator of KLF5. These findings demonstrate that ML264, or an analogue, may hold a promise as a novel therapeutic agent to curb the development and progression of colorectal cancer.
Sigma-Aldrich
ML264, ≥98% (HPLC)