Merck
  • Antioxidant effect of 1,3,4-thiadiazolium mesoionic derivatives on isolated mitochondria.
Antioxidant effect of 1,3,4-thiadiazolium mesoionic derivatives on isolated mitochondria.
European journal of pharmacology (2015-12-17)
Amanda do Rocio Andrade Pires, Gustavo Jabor Gozzi, Guilhermina Rodrigues Noleto, Aurea Echevarria, Camilla Moretto Reis, Maria Eliane Merlin Rocha, Glaucia Regina Martinez, Silvia Maria Suter Correia Cadena
ABSTRACT
Mesoionic compounds have shown antitumor and citotoxic activity against different tumor cells lines, which has been attributed to their physical and chemical characteristics. Among these compounds, the 1,3,4-thiadiazolium-2-phenylamine derivatives have been highlighted due to their important anti-melanoma activity. In this work, the effects of three derivatives that belong this class, MI-J, MI-4F and MI-2,4diF, on the oxidative stress parameters were evaluated using rat liver mitochondria. All the derivatives prevented natural and calcium induced oxidation of pyridine nucleotides at lower concentrations (6.5 and 32.5nmol/mg protein). The calcium uptake was inhibited by all the derivatives at higher concentrations (65 and 130nmol/mg protein), whereas the cation efflux was inhibited only by the MI-J (52%) and MI-4F (50%), possibly by inhibiting the formation of the permeability transition pore (PTP) by 100% and 50%, respectively, as observed in the same experimental conditions. MI-2,4diF did not inhibit the mitochondrial permeability transition or calcium efflux. The enzymatic activity of glutathione reductase, glutathione peroxidase and catalase was not affected by any derivative, but superoxide dismutase was inhibited by all the derivatives. MI-J inhibited enzyme activity significantly (85%) at the highest concentration (130nmol/mg protein); on the other hand, their activity was less affected by fluorine derivatives (MI-4F-20% and MI-2,4diF-32%). These results suggest that these derivatives exert antioxidant effects on isolated mitochondria.
Sigma-Aldrich
MI-4F, ≥98% (HPLC)
Pricing and availability is not currently available.