Merck
  • Stimulatory effect of exogenous diadenosine tetraphosphate on insulin and glucagon secretion in the perfused rat pancreas.
Stimulatory effect of exogenous diadenosine tetraphosphate on insulin and glucagon secretion in the perfused rat pancreas.
British journal of pharmacology (1999-10-12)
R A Silvestre, J Rodríguez-Gallardo, E M Egido, J Marco
ABSTRACT
1. Diadenosine triphosphate (AP3A) and diadenosine tetraphosphate (AP4A) are released by various cells (e.g. platelets and chromaffin cells), and may act as extracellular messengers. In pancreatic B-cells, AP3A and AP4A are inhibitors of the ATP-regulated K+ channels, and glucose increases intracellular levels of both substances. 2. We have studied the effect of exogenous AP3A and AP4A on insulin and glucagon secretion by the perfused rat pancreas. 3. AP3A did not significantly modify insulin or glucagon release, whereas AP4A induced a prompt, short-lived insulin response ( approximately 4 fold higher than basal value; P<0.05) in pancreases perfused at different glucose concentrations (3.2, 5.5 or 9 mM). AP4A-induced insulin release was abolished by somatostatin and by diazoxide. These two substances share the capacity to activate ATP-dependent K+ channels, suggesting that these channels are a potential target for AP4A in the B-cell. 4. AP4A stimulated glucagon release at both 3.2 and 5.5 mM glucose. This effect was abolished by somatostatin. 5. The results suggest that extracellular AP4A may play a physiological role in the control of insulin and glucagon secretion.