Merck
  • Asymmetric nickel-catalyzed Negishi cross-couplings of secondary alpha-bromo amides with organozinc reagents.
Asymmetric nickel-catalyzed Negishi cross-couplings of secondary alpha-bromo amides with organozinc reagents.
Journal of the American Chemical Society (2005-03-31)
Christian Fischer, Gregory C Fu
ABSTRACT
A Ni/Pybox catalyst achieves the asymmetric cross-coupling of secondary alpha-bromo amides with organozinc reagents. The process tolerates a variety of functional groups and affords the desired product in good yield and in high enantiomeric excess.
Sigma-Aldrich
N-Benzyl-2-bromo-N-phenylpropionamide, 97%