Merck
  • MiR-154 directly suppresses DKK2 to activate Wnt signaling pathway and enhance activation of cardiac fibroblasts.
MiR-154 directly suppresses DKK2 to activate Wnt signaling pathway and enhance activation of cardiac fibroblasts.
Cell biology international (2016-08-21)
Li-Ye Sun, Zi-Dong Bie, Chuan-Huan Zhang, Hua Li, Liu-Dong Li, Jun Yang
ABSTRACT
Excessive proliferation of cardiac fibroblasts (CFs) and their transdifferentiation into myofibroblasts leads to expression of α-smooth muscle actin (α-SMA), as well as excessive synthesis and secretion of collagens. This process represents an important pathological basis for myocardial fibrosis (MF). MicroRNA (miR)-154 and the Wnt signaling pathway play key roles in the above process, although their specific interactions are poorly understood. After transfecting CFs with miR-154 mimics or inhibitors, miR-154 was found to inhibit the expression of Dickkopf-related protein 2 (DKK2), while miR-154 inhibitors upregulated DKK2 expression in a Western blot analysis. In a subsequent dual-luciferase activity assay, direct binding of miR-154 to DKK2 was detected. Further experiments demonstrated that transfection of DKK2 siRNA or miR-154 resulted in increased levels of β-catenin, α-SMA, and collagens I and III. Moreover, these changes were observed in association with increases in CF proliferation and migration, and reduced apoptosis. Conversely, transfection of miR-154 inhibitors or DKK2 overexpression vector resulted in lower expression levels of β-catenin, α-SMA, and collagens I and III, suppressed cell proliferation and migration, and enhanced apoptosis. Furthermore, in each assay, when the DKK2 overexpression vector and miR-154 mimics were co-transfected, the functions of each component were counteracted by the other. Therefore, in CFs, targeting of DKK2 by miR-154 leads to upregulation of β-catenin expression and activation of the classical Wnt signaling pathway and CFs. These results suggest new targets for the clinical treatment of MF and ischemic heart disease.