Merck
  • Inhibition of Mcl-1 enhances Pevonedistat-triggered apoptosis in osteosarcoma cells.
Inhibition of Mcl-1 enhances Pevonedistat-triggered apoptosis in osteosarcoma cells.
Experimental cell research (2017-07-01)
Yi Zhang, Chengcheng Shi, Li Yin, Wei Zhou, Haitao Wang, Jingjing Seng, Wencai Li
ABSTRACT
Neddylation inhibitor Pevonedistat (MLN4924) is a novel anticancer drug and has demonstrated broad-spectrum anticancer activity. Nevertheless, we found that Pevonedistat had only a modest apoptotic effect in osteosarcoma (OS) cells. Moreover, we noted that inhibition of neddylation by Pevonedistat led to accumulation of Mcl-1 protein in OS cells. Because Mcl-1 is an important anti-apoptotic protein and also because apoptosis has been shown to be a major cell death pathway, we hypothesized that Mcl-1 accumulation negatively impacted Pevonedistat-mediated anticancer activity in OS cells. In this regard, we employed genetic or pharmacological approaches to inhibit Mcl-1 expression and to examine the effect on Pevonedistat-induced apoptosis in OS cells. We found that inhibition of Mcl-1 expression by siRNA considerably enhanced Pevonedistat-triggered the activation of caspase-3, PARP cleavage and apoptosis, and also dramatically promoted the ability of Pevonedistat to inhibit colony formation of OS cells. Moreover, we observed that flavopiridol, a FDA approved drug, inhibited Mcl-1 expression and substantially enhanced Pevonedistat-mediated activation of apoptosis signaling and significantly augmented cell killing effect in OS cells. Altogether, our study shows that Mcl-1 is a critical resistance factor to Pevonedistat monotherapy, and suggests that Pevonedistat in combinations with flavopiridol may achieve better anticancer therapy.