- Light-induced retinal degeneration causes a transient downregulation of melanopsin in the rat retina.
Light-induced retinal degeneration causes a transient downregulation of melanopsin in the rat retina.
Experimental eye research (2017-05-30)
Diego García-Ayuso, Caridad Galindo-Romero, Johnny Di Pierdomenico, Manuel Vidal-Sanz, Marta Agudo-Barriuso, María P Villegas Pérez
PMID28552384
ABSTRACT
In this work we study the effects of an acute light-induced retinal degeneration on the population of melanopsin positive retinal ganglion cells (m