Merck
  • Effects of fusarium infection on the phenolics in emmer and naked barley.
Effects of fusarium infection on the phenolics in emmer and naked barley.
Journal of agricultural and food chemistry (2010-02-11)
Kai Eggert, Jürgen Hollmann, Beate Hiller, Hans-Peter Kruse, Hashadrai M Rawel, Elke Pawelzik
ABSTRACT
Inoculated or non-inoculated naked barley and emmer cultivars were investigated with regard to their influence on phenolic acid profiles and their arabinoxylan content. Two groups of phenolic compounds were differentiated-methanol-soluble and hydrolyzable covalent-bound phenolic compounds. Chromatographic methods were applied for their analysis. The results showed ferulic acid as the predominant phenol in both total and covalent-bound fractions. The inoculation significantly reduced the ferulic acid content within a range of 5.6-6.6% in the two cereals and all their cultivars. Naked barley cultivars additionally contained the flavonoid catechin in the soluble fraction. The innoculation led here to a significant increase in the catechin content of about 4.5%. These results document an induction of the synthesis of catechin in naked barley after artificial Fusarium infection, whereas the ferulic acid content declined.
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~30 U/mg
Sigma-Aldrich
Cellulase from Aspergillus niger, powder, off-white, ~0.8 U/mg