Merck
P2Y
Cellular and molecular life sciences : CMLS (2016-09-22)
Ramasri Sathanoori, Paulina Bryl-Gorecka, Christa E Müller, Laurie Erb, Gary A Weisman, Björn Olde, David Erlinge
ABSTRACT
Endothelial cells release ATP in response to fluid shear stress, which activates purinergic (P2) receptor-mediated signaling molecules including endothelial nitric oxide (eNOS), a regulator of vascular tone. While P2 receptor-mediated signaling in the vasculature is well studied, the role of P2Y