- A novel chalcone derivative, LQFM064, induces breast cancer cells death via p53, p21, KIT and PDGFRA.
A novel chalcone derivative, LQFM064, induces breast cancer cells death via p53, p21, KIT and PDGFRA.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2017-06-20)
Bruna Lannuce Silva Cabral, Artur Christian Garcia da Silva, Renato Ivan de Ávila, Alane Pereira Cortez, Rangel Magalhães Luzin, Luciano Morais Lião, Eric de Souza Gil, Gérman Sanz, Boniek G Vaz, José R Sabino, Ricardo Menegatti, Marize Campos Valadares
PMID28627468
ABSTRACT
This study shows the design, synthesis and antitumoral potential evaluation of a novel chalcone-like compound, (E)-3- (3, 5-di-ter-butyl-4-hydroxyphenyl)-1- (4-hydroxy-3-methoxyphenyl) prop-2-en-1-one [LQFM064) (4)], against human breast adenocarcinoma MCF7 cells. Some toxicological parameters were also investigated. LQFM064) (4) exhibited cytotoxic activity against MCF7 cells (IC