Merck
  • Small-Molecule-Induced Soluble Oligomers of α-Synuclein with Helical Structure.
Small-Molecule-Induced Soluble Oligomers of α-Synuclein with Helical Structure.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-08-02)
Luis Fonseca-Ornelas, Carla Schmidt, Aldo R Camacho-Zarco, Claudio O Fernandez, Stefan Becker, Markus Zweckstetter
ABSTRACT
Accumulation of α-synuclein (αSyn) aggregates constitutes the hallmark of synucleinopathies including Parkinson's disease. However, many steps from the innocuous, monomeric αSyn toward misfolded oligomers and fibrillar species remain unclear. Here, we show that αSyn can form in solution α-helical oligomers, which are off-pathway to fibrillization, through interaction with the tetrapyrrole phthalocyanine tetrasulfonate. Chemical cross-linking combined with mass spectrometry reveals a large number of intermolecular cross-links along the entire αSyn sequence in the phthalocyanine tetrasulfonate-stabilized αSyn oligomers. Our study suggests that stabilization of structured oligomers by small molecules provides a viable strategy to interfere with αSyn fibrillization.
Sigma-Aldrich
Phthalocyanine tetrasulfonate hydrate, solid
Sigma-Aldrich
Suberic acid bis(3-sulfo-N-hydroxysuccinimide ester) sodium salt, ≥95% (H-NMR), powder