Merck
Bottom-up precise synthesis of stable platinum dimers on graphene.
Nature communications (2017-10-24)
Huan Yan, Yue Lin, Hong Wu, Wenhua Zhang, Zhihu Sun, Hao Cheng, Wei Liu, Chunlei Wang, Junjie Li, Xiaohui Huang, Tao Yao, Jinlong Yang, Shiqiang Wei, Junling Lu
ABSTRACT
Supported metal clusters containing only a few atoms are of great interest. Progress has been made in synthesis of metal single-atom catalysts. However, precise synthesis of metal dimers on high-surface area support remains a grand challenge. Here, we show that Pt
Sigma-Aldrich
Borane-ammonia complex, 95%
Pricing and availability is not currently available.