- The bromodomain and extraterminal domain inhibitor bromosporine synergistically reactivates latent HIV-1 in latently infected cells.
The bromodomain and extraterminal domain inhibitor bromosporine synergistically reactivates latent HIV-1 in latently infected cells.
Oncotarget (2017-12-08)
Hanyu Pan, Panpan Lu, Yinzhong Shen, Yanan Wang, Zhengtao Jiang, Xinyi Yang, Yangcheng Zhong, He Yang, Inam Ulla Khan, Muya Zhou, Bokang Li, Ziyu Zhang, Jianqing Xu, Hongzhou Lu, Huanzhang Zhu
PMID29212213
ABSTRACT
The long-lived latent HIV-1 reservoir is the major barrier for complete cure of Acquired Immune Deficiency Syndrome (AIDS). Here we report that a novel bromodomain and extraterminal domain (BET) inhibitor bromosporine which can broadly target BETs, is able to potently reactivate HIV-1 replication in different latency models alone and more powerful when combined with prostratin or TNF-α. Furthermore, the treatment with bromosporine induced HIV-1 full-length transcripts in resting CD4+ T cells from infected individuals with suppressive antiretroviral therapy (ART)