Merck
  • Optimization of tenofovir release from mucoadhesive vaginal tablets by polymer combination to prevent sexual transmission of HIV.
Optimization of tenofovir release from mucoadhesive vaginal tablets by polymer combination to prevent sexual transmission of HIV.
Carbohydrate polymers (2017-11-08)
Fernando Notario-Pérez, Raúl Cazorla-Luna, Araceli Martín-Illana, Roberto Ruiz-Caro, Aitana Tamayo, Juan Rubio, María-Dolores Veiga
ABSTRACT
The use of sustained-release mucoadhesive vaginal tablets of antiretroviral drugs as microbicidal formulations can be an effective strategy for reducing the sexual transmission of HIV from men to women, which is a main problem particularly in low- and middle-income countries. Different polymers (hydroxypropylmethyl cellulose (HPMC), chitosan, guar gum and Eudragit
Sigma-Aldrich
Calcium phosphate dibasic, puriss. p.a., anhydrous, >98.5% (KT)