Merck
  • High throughput screening of phenoxy carboxylic acids with dispersive solid phase extraction followed by direct analysis in real time mass spectrometry.
High throughput screening of phenoxy carboxylic acids with dispersive solid phase extraction followed by direct analysis in real time mass spectrometry.
Analytica chimica acta (2017-11-16)
Jiaqin Wang, Jun Zhu, Ling Si, Qi Du, Hongli Li, Wentao Bi, David Da Yong Chen
ABSTRACT
A high throughput, low environmental impact methodology for rapid determination of phenoxy carboxylic acids (PCAs) in water samples was developed by combing dispersive solid phase extraction (DSPE) using velvet-like graphitic carbon nitride (V-g-C
Sigma-Aldrich
2,4-Dichlorophenylacetic acid, 99%
Pricing and availability is not currently available.