Merck
  • Long-term treatment with the ghrelin receptor antagonist [d-Lys3]-GHRP-6 does not improve glucose homeostasis in nonobese diabetic MKR mice.
Long-term treatment with the ghrelin receptor antagonist [d-Lys3]-GHRP-6 does not improve glucose homeostasis in nonobese diabetic MKR mice.
American journal of physiology. Regulatory, integrative and comparative physiology (2017-09-15)
Rasha Mosa, Lili Huang, Hongzhuo Li, Michael Grist, Derek LeRoith, Chen Chen
ABSTRACT
Long-term treatment with the ghrelin receptor antagonist [d-Lys3]-GHRP-6 does not improve glucose homeostasis in nonobese diabetic MKR mice. Am J Physiol Regul Integr Comp Physiol 314: R71-R83, 2018. First published September 13, 2017; doi: 10.1152/ajpregu.00157.2017 .-Ghrelin secretion has been associated with increased caloric intake and adiposity. The expressions of ghrelin and its receptor (GHS-R1a) in the pancreas has raised the interest about the role of ghrelin in glucose homeostasis. Most of the studies showed that ghrelin promoted hyperglycemia and inhibited insulin secretion. This raised the interest in using GHS-R1a antagonists as therapeutic targets for type 2 diabetes. Available data of GHS-R antagonists are on a short-term basis. Moreover, the complexity of GHS-R1a signaling makes it difficult to understand the mechanism of action of GHS-R1a antagonists. This study examined the possible effects of long-term treatment with a GHS-R1a antagonist, [d-Lys3]-growth hormone-releasing peptide (GHRP)-6, on glucose homeostasis, food intake, and indirect calorimetric parameters in nonobese diabetic MKR mice. Our results showed that [d-Lys3]-GHRP-6 (200 nmol/mouse) reduced pulsatile growth hormone secretion and body fat mass as expected but worsened glucose and insulin intolerances and increased cumulative food intake unexpectedly. In addition, a significant increase in blood glucose and decreases in plasma insulin and C-peptide levels were observed in MKR mice following long-term [d-Lys3]-GHRP-6 treatment, suggesting a direct inhibition of insulin secretion. Immunofluorescence staining of pancreatic islets showed a proportional increase in somatostatin-positive cells and a decrease in insulin-positive cells in [d-Lys3]-GHRP-6-treated mice. Furthermore, [d-Lys3]-GHRP-6 stimulated food intake on long-term treatment via reduction of proopiomelanocortin gene expression and antagonized GH secretion via reduced growth hormone-releasing hormone gene expression in hypothalamus. These results demonstrate that [d-Lys3]-GHRP-6 is not completely opposite to ghrelin and may not be a treatment option for type 2 diabetes.
Sigma-Aldrich
Free Glycerol Reagent, used for quantitative enzymatic determination of glycerol