Merck
  • Pd-PEPPSI-IPent: an active, sterically demanding cross-coupling catalyst and its application in the synthesis of tetra-ortho-substituted biaryls.
Pd-PEPPSI-IPent: an active, sterically demanding cross-coupling catalyst and its application in the synthesis of tetra-ortho-substituted biaryls.
Angewandte Chemie (International ed. in English) (2009-02-20)
Michael G Organ, Selçuk Calimsiz, Mahmoud Sayah, Ka Hou Hoi, Alan J Lough
ABSTRACT
Incredible Bulk: A series of N-heterocyclic carbene catalysts (see picture) were prepared and evaluated in the Suzuki-Miyaura reaction. A variety of sterically encumbered tetra-ortho-substituted biaryl products were formed from unreactive aryl chlorides using the isopentyl-substituted catalyst at temperatures ranging from 65 degrees C to room temperature. The cyclopentyl-substituted catalyst was virtually inactive, demonstrating that "flexible bulk" is essential to promote these transformations.
Sigma-Aldrich
Pd-PEPPSI-IPent catalyst, ≥95%
Pricing and availability is not currently available.