Merck
Remodeling of phosphatidylglycerol in Synechocystis PCC6803.
Biochimica et biophysica acta (2009-10-28)
Hajnalka Laczko-Dobos, Petr Frycák, Bettina Ughy, Ildiko Domonkos, Hajime Wada, Laszlo Prokai, Zoltan Gombos
ABSTRACT
The phosphatidylglycerol deficient DeltapgsA mutant of Synechocystis PCC6803 provided a unique experimental system for investigating in vivo retailoring of exogenously added dioleoylphosphatidylglycerol in phosphatidylglycerol-depleted cells. Gas chromatographic analysis of fatty acid composition suggested that diacyl-phosphatidylglycerols were synthesized from the artificial synthetic precursor. The formation of new, retailored lipid species was confirmed by negative-ion electrospray ionization-Fourier-transform ion cyclotron resonance and ion trap tandem mass spectrometry. Various isomeric diacyl-phosphatidylglycerols were identified indicating transesterification of the exogenously added dioleoylphosphatidyl-glycerol at the sn-1 or sn-2 positions. Polyunsaturated fatty acids were incorporated selectively into the sn-1 position. Our experiments with Synechocystis PCC6803/DeltapgsA mutant cells demonstrated lipid remodeling in a prokaryotic photosynthetic bacterium. Our data suggest that the remodeling of diacylphosphatidylglycerol likely involves reactions catalyzed by phospholipase A(1) and A(2) or acyl-hydrolase, lysophosphatidylglycerol acyltransferase and acyl-lipid desaturases.
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt, ≥98.0% (TLC)