Skip to Content
Merck

AABH97D2C2D8

AA BLOCKS, INC.

Arsenobetaine

Synonym(s):

(trimethylarsonio)acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H11O2As1
CAS Number:
Molecular Weight:
178.06
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H11AsO2/c1-6(2,3)4-5(7)8/h4H2,1-3H3

Inchi Key

SPTHHTGLGVZZRH-UHFFFAOYSA-N

Purity

95%

Storage temp.

N/A

Country of Origin

US



Still not finding the right product?

Explore all of our products under Arsenobetaine




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library