Skip to Content
Merck

AABH99A42A5E

AA BLOCKS, INC.

Ferroin Sulfate (0.025M aq solution) (Technical Grade)

Synonym(s):

iron(2+) sulfate compound with [1,10]phenanthroline (1:3)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C36H24O4N6S1Fe1
CAS Number:
Molecular Weight:
692.54
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/3C12H8N2.Fe.H2O4S/c3*1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1;;1-5(2,3)4/h3*1-8H;;(H2,1,2,3,4)/q;;;+2;/p-2

Inchi Key

CIWXFRVOSDNDJZ-UHFFFAOYSA-L

Storage temp.

RT

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library