biological source
hamster ovary (CHO cells)
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin (Purified animal free recombinant blocking antibody)
clone
polyclonal
description
Blocking Antibody (Recombinant Anti-Human IgG Fc fragment)
form
liquid ( in PBS buffer containing 0.09% sodium azide)
species reactivity
human (Anti-Human IgG Fc Fragment)
packaging
PVC-coated bottle of 15 g
concentration
≥95%
technique(s)
immunoassay: suitable (Various)
isotype
IgG
shipped in
2-8°C
storage temp.
2-8°C
General description
SuperFc Block - Recombinant Anti-Human IgG, Fc fragment is an animal-free recombinant blocking antibody optimized for use in an antibody-based immunoassay to remove disruptive antibodies in serum, such as human anti-mouse antibodies (HAMA) and rheumatoid factor (RF).
SuperFc Block inhibits heterophilic antibodies like HAMA or RF. By binding to these unwanted antibodies it efficiently reduces non-specific antibody interference from contributing to false positive and negative test results, that require superior consistency and performance. The recombinant Anti-Human IgG, Fc fragment blocker is an animal-free, sustainably sourced material that supports ethical research practices.
SuperFc Block inhibits heterophilic antibodies like HAMA or RF. By binding to these unwanted antibodies it efficiently reduces non-specific antibody interference from contributing to false positive and negative test results, that require superior consistency and performance. The recombinant Anti-Human IgG, Fc fragment blocker is an animal-free, sustainably sourced material that supports ethical research practices.
Application
- SuperFc Block is a critical reagent that inhibits heterophilic antibodies like HAMA or RF by binding unwanted antibodies to efficiently reduces non-specific antibody interference that could be contributing to false positive and negative test results.
- SuperFc Block is suitable for use with lateral flow, CLIA and ELISA tests including competitive, sandwich, and multiplex detection assays.
- As a recombinant Anti-Human IgG, Fc fragment blocker, it is animal-free, a sustainable sourced antibody that limits variability between batches, ensuring consistent performance.
Features and Benefits
- Cell Culture, Clonal-Derived Material, limits variability between batches, ensuring consistent performance.
- Animal-Free Production provides sustainably sourced material that supports ethical research practices.
- Elimination of Pooled Serum, guarantees consistent purity and eliminates animal herd variability.
- Less Complex Material, reduces interfering substances inherent in serum, leading to more accurate results.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service