Merck
  • ROS-producing immature neutrophils in giant cell arteritis are linked to vascular pathologies.
ROS-producing immature neutrophils in giant cell arteritis are linked to vascular pathologies.
JCI insight (2020-09-23)
Lihui Wang, Zhichao Ai, Tariq Khoyratty, Kristina Zec, Hayley L Eames, Erinke van Grinsven, Alison Hudak, Susan Morris, David Ahern, Claudia Monaco, Evgeniy B Eruslanov, Raashid Luqmani, Irina A Udalova
ABSTRACT
Giant cell arteritis (GCA) is a common form of primary systemic vasculitis in adults, with no reliable indicators of prognosis or treatment responses. We used single cell technologies to comprehensively map immune cell populations in the blood of patients with GCA and identified the CD66b+CD15+CD10lo/-CD64- band neutrophils and CD66bhiCD15+CD10lo/-CD64+/bright myelocytes/metamyelocytes to be unequivocally associated with both the clinical phenotype and response to treatment. Immature neutrophils were resistant to apoptosis, remained in the vasculature for a prolonged period of time, interacted with platelets, and extravasated into the tissue surrounding the temporal arteries of patients with GCA. We discovered that immature neutrophils generated high levels of extracellular reactive oxygen species, leading to enhanced protein oxidation and permeability of endothelial barrier in an in vitro coculture system. The same populations were also detected in other systemic vasculitides. These findings link functions of immature neutrophils to disease pathogenesis, establishing a clinical cellular signature of GCA and suggesting different therapeutic approaches in systemic vascular inflammation.
Sigma-Aldrich
Dihydrorhodamine 123, ≥95%
Sigma-Aldrich
In Vitro Vascular Permeability Assay (24-well), This In Vitro Vascular Permeability Assay kit employs a 24-well plate and provides an efficient system for evaluating the effects of chemicals & drug compounds on endothelial cell adsorption, transport & permeability.