Skip to Content
MilliporeSigma

COMH04239653

Combi-Blocks, Inc.

5-Methyl-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid

Synonym(s):

5-methyl-2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H6O4N2
CAS Number:
Molecular Weight:
170.12
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H6N2O4/c1-2-3(5(10)11)7-6(12)8-4(2)9/h1H3,(H,10,11)(H2,7,8,9,12)

Inchi Key

UQBMFARVSOQVQE-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library