Product Name
4-Methoxy-3-methylbenzaldehyde 99%
Product Number
152129
Product Brand
ALDRICH
CAS Number
32723-67-4
Molecular Weight
150.17
Test
Specification
APPEARANCE (COLOR)Colorless to Light Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 98.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE