Product Name
Tetrachloro-1,4-benzoquinone 99%
Product Number
232017
Product Brand
ALDRICH
CAS Number
118-75-2
Molecular Weight
245.88
Test
Specification
APPEARANCE (COLOR)Yellow to Yellow-Green
APPEARANCE (FORM)Powder or Crystals
TITRATION98.5 - 101.5 %
TITRATION (METHOD)WITH AGNO3 AFTER OXYGEN COMBUSTION
PURITY (HPLC AREA %)≥ 98.5 %
SOLUBILITY (COLOR)Yellow to Yellow-Green
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)3.3MG/10ML CHLOROFORM
INFRARED SPECTRUMCONFORMS TO STRUCTURE