Product Name
3-Nitrophthalic acid 95%
Product Number
369306
Product Brand
ALDRICH
CAS Number
603-11-2
Molecular Weight
211.13
Test | Specification |
|---|---|
| APPEARANCE (COLOR) | White to Off White |
| APPEARANCE (FORM) | Powder or Crystals |
| TITRATION (T) NAOH 0.1M | 94.5 - 105.5 % |
| PURITY (HPLC AREA %) | ≥ 94.5 % |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |