Product Name
5-(4-Bromophenyl)oxazole-2-propionic acid 97%
Product Number
658561
Product Brand
ALDRICH
CAS Number
23464-96-2
Molecular Weight
296.12
Test
Specification
APPEARANCEWHITE TO BROWN POWDER, CRYSTALS OR
 CRYSTALLINE POWDER AND/OR CHUNKS
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
TITRATION96.5% - 103.5% (WITH NAOH)
GAS LIQUID96.5% (MINIMUM)
 APPROVED AUGUST 30, 2005 BTR