Product Name
2-Hexanone analytical standard
Product Number
02473
Product Brand
SIAL
CAS Number
591-78-6
Molecular Weight
100.16
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 99.5 %
REFRACTIVE INDEX N20/D1.400 - 1.402
WATER (COULOMETR.)≤ 0.2 %
EXPIRY PERIOD36 MONTHS