Product Name
Alkane standard solution C8-C20 analytical standard, contains C8-C20, ~40 mg/L each, in hexane
Product Number
04070
Product Brand
SIAL
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)CONTAINS 13 COMPOUNDS (C8 - C20)
DENSITY D20/40.659 - 0.661
REFRACTIVE INDEX N20/D1.374 - 1.376