Product Name
O-Succinyl-L-carnitine lithium salt ≥98.0% (TLC)
Product Number
04609
Product Brand
SIAL
CAS Number
Molecular Weight
261.27 (free acid basis)
Test
Specification
Assay (QNMR)*Report result (calculated on organic component)
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (TLC AREA %)≥ 98.0 %
SPECIFIC ROTATION (20/D)-16.0 ± 3.0 Degrees
CONCENTRATIONC=0.1 IN WATER
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
CARBON 13 NMR SPECTRUMCONFORMS TO STRUCTURE
AAS QUALITATIVELithium confirmed