Product Name
Ammonium bicarbonate BioUltra, ≥99.5% (T)
Product Number
09830
Product Brand
SIAL
CAS Number
1066-33-7
Molecular Weight
79.06
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder or Crystals
TITRATION (T) HCL 1M99.5 - 101.0 %
RESIDUE ON IGNITION≤ 0.05 %
METAL TRACE ANALYSIS (ICP)CORRESPONDS TO REQUIREMENTS
ALUMINIUM (ICP)≤ 5 mg/kg
BARIUM (ICP)≤ 5 mg/kg
BISMUTH (ICP)≤ 5 mg/kg
CALCIUM (ICP)≤ 10 mg/kg
CADMIUM (ICP)≤ 5 mg/kg
COBALT (ICP)≤ 5 mg/kg
CHROMIUM (ICP)≤ 5 mg/kg
COPPER (ICP)≤ 5 mg/kg
IRON (ICP)≤ 5 mg/kg
POTASSIUM (ICP)≤ 50 mg/kg
LITHIUM (ICP)≤ 5 mg/kg
MAGNESIUM (ICP)≤ 500 mg/kg
MANGANESE (ICP)≤ 5 mg/kg
MOLYBDENUM (ICP)≤ 5 mg/kg
SODIUM (ICP)≤ 50 mg/kg
NICKEL (ICP)≤ 5 mg/kg
LEAD (ICP)≤ 5 mg/kg
STRONTIUM (ICP)≤ 5 mg/kg
ZINC (ICP)≤ 5 mg/kg
ARSENIC TRACES (MHS-AAS)≤ 0.1 mg/kg
TOTAL SULFUR AS SO4 (ICP)≤ 50 mg/kg
CHLORIDE (CL)≤ 50 mg/kg
SOLUBILITY (METHOD)1 M IN WATER
APPEARANCE (SOLUTION)CLEAR COLORLESS
PH (SOLUTION)7.0 - 8.5
RESIDUE (FILTER TEST)NO RESIDUE
UV - ABS. AT 260 NM≤ 0.030
UV - ABS. AT 280 NM≤ 0.020
RECOMMENDED RETEST PERIOD72 MONTHS