Product Name
Silica gel orange granular, 0.2-1 mm
Product Number
10087
Product Brand
SIAL
CAS Number
112926-00-8
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Orange
APPEARANCE (FORM)Granules
PARTICLE SIZECORRESPONDS TO REQUIREMENTS
LOSS ON DRYING≤ 2.0 %
APPLICATION TESTWATER ADSORPTION CAPACITY
>30%, (72H)
PROOF OF IDENTITYINDICATOR REACTION: CORRESPONDS
RECOMMENDED RETEST PERIOD36 MONTHS