Product Name
Oleoyl-L-carnitine analytical standard
Product Number
19945
Product Brand
SIAL
CAS Number
38677-66-6
Molecular Weight
425.64
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 95.0 %
IDENTIFICATION (LC-MS)CORRESPONDS TO REQUIREMENTS
SPECIFIC ROTATION (20/D)-13.0 ± 2.0 Degrees
CONCENTRATIONc=0.1 in methanol
REM. ON ELEMENTAL ANALYSIS≤ 10 % water (calcd. from elemental analysis)
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS