Product Name
Arachidonyl-L-carnitine analytical standard
Product Number
30172
Product Brand
SIAL
CAS Number
36816-11-2
Molecular Weight
447.65
Test
Specification
APPEARANCE (COLOR)White to Very Dark Yellow to Very Dark Red
APPEARANCE (FORM)Powder or Solid or Crystals
PURITY (HPLC AREA %)≥ 95.0 %
SPECIFIC ROTATION (20/D)-16.0 - -10.0 Degrees
CONCENTRATIONc=0.1 in methanol
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
CARBON 13 NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS