Product Name
Pyrasulfotole PESTANAL®, analytical standard
Product Number
32973
Product Brand
SIAL
CAS Number
365400-11-9
Molecular Weight
362.32
Test
Specification
APPEARANCE (COLOR)White to Light Yellow to Light Beige
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
MELTING POINT199 - 205 C
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS