Product Name
Cyazofamid PESTANAL®, analytical standard
Product Number
33874
Product Brand
SIAL
CAS Number
120116-88-3
Molecular Weight
324.79
Test
Specification
APPEARANCE (COLOR)White to Light Yellow to Light Beige
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
MELTING POINT155 - 161 C
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS