Product Name
Diallyl phthalate analytical standard
Product Number
36925
Product Brand
SIAL
CAS Number
131-17-9
Molecular Weight
246.26
Test
Specification
APPEARANCE (COLOR)Colorless to Green-Yellow to Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 98.0 %
REFRACTIVE INDEX N20/D1.5177 - 1.5217
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS