Product Name
trans-4-Hydroxy-L-proline analytical standard
Product Number
41875
Product Brand
SIAL
CAS Number
51-35-4
Molecular Weight
131.13
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder or Crystals
PURITY (TLC AREA %)≥ 99 %
SPECIFIC ROTATION (20/D)-76.0 ± 1.5 Degrees
CONCENTRATIONC=5 IN WATER
INFRARED SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS