Product Name
Flurtamone PESTANAL®, analytical standard
Product Number
46286
Product Brand
SIAL
CAS Number
96525-23-4
Molecular Weight
333.30
Test
Specification
APPEARANCE (COLOR)White to Light Yellow to Light Beige
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
MELTING POINT152 - 157 C
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS