Product Name
Fipronil PESTANAL®, analytical standard
Product Number
46451
Product Brand
SIAL
CAS Number
120068-37-3
Molecular Weight
437.15
Test | Specification |
|---|---|
| APPEARANCE (COLOR) | White to Faint Yellow to Faint Beige |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (HPLC AREA %) | ≥ 95.0 % |
| MELTING POINT | 195 - 205 C |
| WATER | ≤ 2.0 % |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 48 MONTHS |