Product Name
(±)-Diclofop PESTANAL®, analytical standard
Product Number
52256
Product Brand
SIAL
CAS Number
40843-25-2
Molecular Weight
327.16
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder
PURITY (HPLC AREA %)≥ 98.0 %
MELTING POINT99 - 107 C
CARBON CONTENT54.00 - 56.20 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
CARBON 13 NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD24 MONTHS