Product Name
Silica gel high-purity grade (w/ Ca, ~0.1%), pore size 60 Å, 230-400 mesh particle size
Product Number
60752
Product Brand
SIAL
CAS Number
112926-00-8
Molecular Weight
Test
Specification
LOSS ON DRYING≤ 9.0 %
CALCIUM0.10 - 0.30 %
RECOMMENDED RETEST PERIOD48 MONTHS