Product Name
7-(Diethylamino)coumarin-3-carboxylic acid BioReagent, suitable for fluorescence, ≥98.0% (HPCE)
Product Number
36799
Product Brand
SIGMA
CAS Number
50995-74-9
Molecular Weight
261.27
Test
Specification
APPEARANCE (COLOR)Faint Orange to Orange
APPEARANCE (FORM)Powder or Crystals
PURITY (HPCE AREA %)≥ 98.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EMISSION MAXIMUM473 ± 5 nm
REMARKS ON LUMINESCENCEEX. 409 NM IN 0.1 M TRIS PH 9.0