Product Name
Boc-Ala-Pro-Nva p-chlorothiobenzyl ester
Product Number
B2017
Product Brand
SIGMA
CAS Number
90171-50-9
Molecular Weight
526.09
Test
Specification
APPEARANCEWHITE POWDER
SOLUBILITYCLEAR COLORLESS SOLUTION AT 10 MG/ ML OF CHLOROFORM
ELEMENTAL ANALYSIS55.6 - 58.5% CARBON 7.7 - 8.3% NITROGEN
PURITY BY THIN LAYER CHROMATOGRAPHY> OR = 98%
DOCUMENT #B2017/05/09/08/1