Product Name
L-Citrulline ≥98% (TLC)
Product Number
C7629
Product Brand
SIGMA
CAS Number
372-75-8
Molecular Weight
175.19
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder
PURITY (TLC AREA %)≥ 98 %
SOLUBILITY (COLOR)Colorless
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)50MG/ML WATER
CARBON CONTENT40.3 - 42.0 %
NITROGEN CONTENT23.5 - 24.4 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST7 YEARS