Product Name
Fluorescein isothiocyanate–dextran average mol wt 3,000-5,000
Product Number
FD4
Product Brand
SIGMA
CAS Number
60842-46-8
Molecular Weight
Test
Specification
DEGREE OF SUBSTITUTION0.001 - 0.020 mol FITC/mol of Glucose
RECOMMENDED RETEST PERIOD24 MONTHS