Product Name
Moxidectin VETRANAL®, analytical standard
Product Number
33746
Product Brand
sial
CAS Number
113507-06-5
Molecular Weight
639.82
Test
Specification
APPEARANCE (COLOR)White to Faint Yellow to Faint Beige
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 95.0 %
MELTING POINT130 - 138 C
WATER≤ 3.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD24 MONTHS