Product Name
trans-2-Tetradecenoyl-L-carnitine analytical standard
Product Number
40733
Product Brand
sial
CAS Number
Molecular Weight
369.54
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (LC-MS)≥ 95.0 %
SPECIFIC ROTATION (20/D)-12 ± 2 Degrees
CONCENTRATIONc=0.1 in methanol
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS